Gene Transcripts

Gene Symbol
C2orf47
Entrez Gene ID
79568
Gene Name
chromosome 2 open reading frame 47
Gene Aliases
Chromosome Location
Chr. 2: 199955317 - 199964125 on Build GRCh38
UniGene ID
Hs.154494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024520.2 NP_078796.2 4 876
XM_005246852.2 XP_005246909.1 3 787
GenBank mRNA AK026208.1 - 4 876
AL832964.1 - 3 195
BC017959.1 - 3 560 4607945

View Full Product Details