Gene Transcripts

Gene Symbol
SLC7A2
Entrez Gene ID
6542
Gene Name
solute carrier family 7 member 2
Gene Aliases
ATRC2, CAT2, HCAT2
Chromosome Location
Chr. 8: 17494349 - 17570568 on Build GRCh38
UniGene ID
Hs.448520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008539.3 NP_001008539.3 3, 4 712
NM_001164771.1 NP_001158243.1 3, 4 708
NM_003046.5 NP_003037.4 3, 4 708
XM_005273610.4 XP_005273667.1 4, 5 773
XM_005273611.4 XP_005273668.1 4, 5 676
XM_005273612.4 XP_005273669.1 4, 5 770
XM_017013746.1 XP_016869235.1 3, 4 711
XM_017013747.1 XP_016869236.1 4, 5 773
GenBank mRNA AK314287.1 - 3, 4 710
BC069648.1 - 3, 4 615 7262473
BC104905.1 - 3, 4 601 8143908
BC113661.1 - 3, 4 601 8322713
BC143583.1 - 3, 4 601 9052094
D29990.1 - 2, 3 540
U76368.1 - 3, 4 734

View Full Product Details