Gene Transcripts
- Gene Symbol
- SPICE1
- Entrez Gene ID
- 152185
- Gene Name
- spindle and centriole associated protein 1
- Gene Aliases
- CCDC52, SPICE
- Chromosome Location
- Chr. 3: 113442718 - 113515187 on Build GRCh38
- UniGene ID
- Hs.477144
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144718.3 |
NP_653319.1
|
5, 6 |
684 |
|
XM_011512461.2 |
XP_011510763.1
|
5, 6 |
441 |
|
XM_017005775.1 |
XP_016861264.1
|
5, 6 |
441 |
|
XM_017005776.1 |
XP_016861265.1
|
5, 6 |
578 |
|
XM_017005777.1 |
XP_016861266.1
|
5, 6 |
501 |
|
XM_017005778.1 |
XP_016861267.1
|
5, 6 |
685 |
|
GenBank mRNA |
AK126897.1 |
- |
1, 2 |
90 |
|
AL832148.1 |
- |
5, 6 |
653 |
|
AL832327.2 |
- |
5, 6 |
467 |
|
AY099107.1 |
- |
5, 6 |
604 |
|
BC036951.1 |
- |
5, 6 |
625 |
5500345 |
View Full Product Details