Gene Transcripts

Gene Symbol
XXYLT1
Entrez Gene ID
152002
Gene Name
xyloside xylosyltransferase 1
Gene Aliases
C3orf21
Chromosome Location
Chr. 3: 195068276 - 195271166 on Build GRCh38
UniGene ID
Hs.478741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308069.1 NP_001294998.1 4 1217
NM_152531.4 NP_689744.3 4 1680
XM_005269286.4 XP_005269343.1 4 4065
XM_011512448.2 XP_011510750.1 5 1492
XM_017005750.1 XP_016861239.1 3 4453
GenBank mRNA AK057046.1 - 5 1425
AK075551.1 - 4 1648
AK092474.1 - 2 957
BC019036.2 - 4 1589 4591786
BC039067.1 - 4 1662 5185557

View Full Product Details