Gene Transcripts

Gene Symbol
BGN
Entrez Gene ID
633
Gene Name
biglycan
Gene Aliases
DSPG1, PG-S1, PGI, SLRR1A
Chromosome Location
Chr. X: 153494889 - 153509554 on Build GRCh38
UniGene ID
Hs.821
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001711.5 NP_001702.1 2 334
XM_017029724.1 XP_016885213.1 1 113
GenBank mRNA AK092954.1 - 2 242
AK093023.1 - 2 240
AK222484.1 - 2 243
AK223302.1 - 2 214
AK291955.1 - 2 245
AK293268.1 - 2 270
AK293289.1 - 2 169
AK297966.1 - 2 243
AK298756.1 - 1 43
AU142113.1 - 2 270
BC002416.2 - 2 227 3162633
BC004244.1 - 2 237 3503374
BT007323.1 - 1 98
DC299956.1 - 2 334
DC419932.1 - 2 314
DC425011.1 - 2 295
J04599.1 - 2 218

View Full Product Details