Gene Transcripts

Gene Symbol
CNKSR2
Entrez Gene ID
22866
Gene Name
connector enhancer of kinase suppressor of Ras 2
Gene Aliases
CNK2, KSR2, MAGUIN
Chromosome Location
Chr. X: 21374335 - 21654695 on Build GRCh38
UniGene ID
Hs.555917
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168648.1 NP_001162119.1 12 1796
NM_001168649.1 NP_001162120.1 11 1649
NM_014927.3 NP_055742.2 12 1796
XM_011545472.2 XP_011543774.1 11 1653
GenBank mRNA AB020709.1 - 12 1772
AF418269.1 - 12 1352
AF418270.1 - 12 1352
AK294728.1 - 11 1611
BC126121.1 - 12 1351 8991837
BC136289.1 - 12 1351 9020276

View Full Product Details