Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016024.2 | NP_057108.2 | 6 | 956 | |
GenBank mRNA | AF151837.1 | - | 5 | 891 | |
AK292855.1 | - | 6 | 938 | ||
AL050405.1 | - | 6 | 931 | ||
BC033750.1 | - | 6 | 924 | 5210341 | |
BC040433.1 | - | 6 | 907 |