Gene Transcripts

Gene Symbol
PRR16
Entrez Gene ID
51334
Gene Name
proline rich 16
Gene Aliases
DSC54, LARGEN
Chromosome Location
Chr. 5: 120464222 - 120794587 on Build GRCh38
UniGene ID
Hs.157461
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300783.1 NP_001287712.1 2 663
NM_001308087.1 NP_001295016.1 2 376
NM_001308088.1 NP_001295017.1 4 616
NM_016644.2 NP_057728.1 3 788
XM_011543452.2 XP_011541754.1 3 2275
XM_011543453.2 XP_011541755.1 3 757
XM_011543454.2 XP_011541756.1 4 863
GenBank mRNA AF242769.1 - 3 742
BC038838.1 - 3 475 5170587
BC092474.1 - 2 664 30528467
BM728431.1 - 4 616
DA328660.1 - 2 377

View Full Product Details