Gene Transcripts
- Gene Symbol
- TPT1P8
- Entrez Gene ID
- 59347
- Gene Name
- tumor protein, translationally-controlled 1 pseudogene 8
- Gene Aliases
- FKSG2
- Chromosome Location
- Chr. 8: 36888391 - 36889522 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF300871.1 |
- |
1 |
202 |
|
View Full Product Details