Gene Transcripts

Gene Symbol
RBM33
Entrez Gene ID
155435
Gene Name
RNA binding motif protein 33
Gene Aliases
PRR8
Chromosome Location
Chr. 7: 155644494 - 155781485 on Build GRCh38
UniGene ID
Hs.591815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053043.2 NP_444271.2 13 2570
XM_005249531.1 XP_005249588.1 14 2735
XM_005249532.1 XP_005249589.1 14 2735
XM_005249533.1 XP_005249590.1 14 2705
XM_005249534.1 XP_005249591.1 13 2615
XM_005249535.1 XP_005249592.1 13 2585
XM_011515840.1 XP_011514142.1 14 2735
GenBank mRNA AK301179.1 - 8 1517
AK307515.1 - 13 2400
AL137724.1 - 11 3465

View Full Product Details