Gene Transcripts

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
Chromosome Location
Chr. 7: 117363222 - 117427523 on Build GRCh38
UniGene ID
Hs.352412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301821.1 NP_001288750.1 6 642
NM_001301822.1 NP_001288751.1 5 530
NM_130768.2 NP_570124.1 6 642
GenBank mRNA AF461259.1 - 6 608
AK093445.1 - 6 595
BC028695.1 - 6 612
BC034963.1 - 6 617 4824818
BC068488.1 - 5 503
BC126186.1 - 6 601 8991902
BC126188.1 - 6 601 8991904
BC144212.1 - 6 601 9052732

View Full Product Details