Gene Transcripts

Gene Symbol
LIX1
Entrez Gene ID
167410
Gene Name
limb and CNS expressed 1
Gene Aliases
C5orf11, Lft
Chromosome Location
Chr. 5: 97091870 - 97142816 on Build GRCh38
UniGene ID
Hs.656702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153234.4 NP_694966.3 6 1575
GenBank mRNA AK098400.1 - 6 1631
BC036467.2 - 6 1494 5264055

View Full Product Details