Gene Transcripts

Gene Symbol
PIP4K2C
Entrez Gene ID
79837
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 gamma
Gene Aliases
PIP5K2C
Chromosome Location
Chr. 12: 57591159 - 57603428 on Build GRCh38
UniGene ID
Hs.745011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146258.1 NP_001139730.1 2 388
NM_001146260.1 NP_001139732.1 2 388
NM_024779.4 NP_079055.3 2 388
XM_005269152.2 XP_005269209.1 2 381
XM_011538747.1 XP_011537049.1 2 381
XM_017019973.1 XP_016875462.1 2 345
GenBank mRNA AK075553.1 - 2 344
AK125526.1 - 2 272
AK297243.1 - 2 373
AK315588.1 - 2 268
AX754970.1 - 2 329
BC028596.2 - 2 273 4824916
BX419431.2 - 2 386

View Full Product Details