Gene Transcripts

Gene Symbol
PRELID2
Entrez Gene ID
153768
Gene Name
PRELI domain containing 2
Gene Aliases
Chromosome Location
Chr. 5: 145587325 - 145835369 on Build GRCh38
UniGene ID
Hs.314261
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138492.5 NP_612501.3 8 1379
NM_182960.3 NP_892005.1 8 1212
NM_205846.2 NP_995318.1 7 1176
GenBank mRNA AK095695.1 - 8 1145

View Full Product Details