Gene Transcripts

Gene Symbol
ARHGAP18
Entrez Gene ID
93663
Gene Name
Rho GTPase activating protein 18
Gene Aliases
MacGAP, SENEX, bA307O14.2
Chromosome Location
Chr. 6: 129576145 - 129710225 on Build GRCh38
UniGene ID
Hs.486458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033515.2 NP_277050.2 12 1702
XM_005267212.2 XP_005267269.2 12 1778
XM_005267213.1 XP_005267270.1 12 1702
XM_017011494.1 XP_016866983.1 12 1778
GenBank mRNA AB053293.1 - 11 1500
AL834511.1 - 12 1640
AY643712.1 - 12 1703
BC101708.1 - 12 1696 8069214
BC107416.1 - 12 1658 4040931
BC111940.1 - 12 1696 8327408
BC143531.1 - 12 1696

View Full Product Details