Gene Transcripts

Gene Symbol
TSGA13
Entrez Gene ID
114960
Gene Name
testis specific 13
Gene Aliases
Chromosome Location
Chr. 7: 130668646 - 130687432 on Build GRCh38
UniGene ID
Hs.592266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304968.1 NP_001291897.1 6 1105
NM_052933.3 NP_443165.1 5 710
GenBank mRNA AF294347.1 - 5 428
AF431815.1 - 5 710
AK093329.1 - 6 1104
BC038415.1 - 5 710 5172129

View Full Product Details