Gene Transcripts
- Gene Symbol
- ACSM1
- Entrez Gene ID
- 116285
- Gene Name
- acyl-CoA synthetase medium-chain family member 1
- Gene Aliases
- BUCS1, MACS1
- Chromosome Location
- Chr. 16: 20623235 - 20697788 on Build GRCh38
- UniGene ID
- Hs.306812
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318890.1 |
NP_001305819.1
|
6, 7 |
1112 |
|
NM_052956.2 |
NP_443188.2
|
5, 6 |
976 |
|
NR_134918.1 |
|
6, 7 |
1037 |
|
XM_006721016.2 |
XP_006721079.1
|
5, 6 |
955 |
|
XM_006721017.2 |
XP_006721080.1
|
5, 6 |
955 |
|
XM_006721018.2 |
XP_006721081.1
|
3, 4 |
334 |
|
XM_011545729.2 |
XP_011544031.1
|
5, 6 |
955 |
|
XM_011545730.2 |
XP_011544032.1
|
5, 6 |
955 |
|
XM_011545731.2 |
XP_011544033.1
|
3, 4 |
1151 |
|
XM_011545732.2 |
XP_011544034.1
|
4, 5 |
393 |
|
XM_017022914.1 |
XP_016878403.1
|
5, 6 |
955 |
|
XM_017022915.1 |
XP_016878404.1
|
4, 5 |
1803 |
|
GenBank mRNA |
AB059429.1 |
- |
5, 6 |
976 |
|
AK092574.1 |
- |
3, 4 |
334 |
|
AK314282.1 |
- |
6, 7 |
1068 |
|
BC125177.1 |
- |
6, 7 |
1037 |
|
BC125178.1 |
- |
5, 6 |
976 |
40121956 |
View Full Product Details