Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099691.2 | NP_001093161.1 | 6 | 2212 | |
NM_001308158.1 | NP_001295087.1 | 6 | 2382 | ||
NM_001308159.1 | NP_001295088.1 | 6 | 2379 | ||
NM_003236.3 | NP_003227.1 | 6 | 2215 | ||
GenBank mRNA | AF149096.1 | - | 6 | 726 | |
AF149097.1 | - | 6 | 723 | ||
AF149098.1 | - | 7 | 755 | ||
BM453873.1 | - | 1 | 319 | ||
DA373586.1 | - | 1 | 273 | ||
X70340.1 | - | 6 | 1997 |