Gene Transcripts

Gene Symbol
SFT2D1
Entrez Gene ID
113402
Gene Name
SFT2 domain containing 1
Gene Aliases
C6orf83, pRGR1
Chromosome Location
Chr. 6: 166319728 - 166342606 on Build GRCh38
UniGene ID
Hs.487143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145169.2 NP_660152.1 8 697
NR_130112.1 9 720
NR_130113.1 7 625
NR_130114.1 7 616
GenBank mRNA AF041429.1 - 8 631
AK021489.1 - 2 1964
AK090795.1 - 1 1604
AL833217.1 - 2 3028
AW964442.1 - 7 547
BC018969.1 - 8 602 3959886
BC028388.1 - 8 2315

View Full Product Details