Gene Transcripts

Gene Symbol
FAM219A
Entrez Gene ID
203259
Gene Name
family with sequence similarity 219 member A
Gene Aliases
C9orf25
Chromosome Location
Chr. 9: 34398184 - 34458610 on Build GRCh38
UniGene ID
Hs.493771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184940.1 NP_001171869.1 6 2560
NM_001184941.1 NP_001171870.1 6 2557
NM_001184942.1 NP_001171871.1 6 2524
NM_001184943.1 NP_001171872.1 6 2506
NM_001184945.1 NP_001171874.1 6 2473
NM_147202.1 NP_671735.1 6 2509
XM_005251402.4 XP_005251459.1 7 2767
XM_011517797.2 XP_011516099.1 6 2567
XM_011517798.2 XP_011516100.1 6 2516
XM_011517799.2 XP_011516101.1 6 2365
GenBank mRNA AF131828.1 - 1 700
AK022819.1 - 1 1178
AK026087.1 - 1 302
AK091262.1 - 2 1899
AK096350.1 - 6 2509
AK123887.1 - 7 2367
AL833397.1 - 1 1506
BC015785.1 - 1 923
BC025348.1 - 2 1917
BC041009.2 - 6 2473 5284905

Gene Symbol
C9orf24
Entrez Gene ID
84688
Gene Name
chromosome 9 open reading frame 24
Gene Aliases
CBE1, NYD-SP22, SMRP1, bA573M23.4
Chromosome Location
Chr. 9: 34379019 - 34400833 on Build GRCh38
UniGene ID
Hs.50334
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005251616.4 XP_005251673.1 1 1565

View Full Product Details