Gene Transcripts

Gene Symbol
TSSK4
Entrez Gene ID
283629
Gene Name
testis specific serine kinase 4
Gene Aliases
C14orf20, STK22E, TSK-4, TSK4, TSSK5
Chromosome Location
Chr. 14: 24205524 - 24208362 on Build GRCh38
UniGene ID
Hs.314432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184739.1 NP_001171668.1 2 493
NM_001308067.1 NP_001294996.1 2 218
NM_174944.3 NP_777604.2 2 493
XM_011536663.2 XP_011534965.1 2 345
XM_017021226.1 XP_016876715.1 2 689
GenBank mRNA AF542390.1 - 1 64
AK131042.1 - 2 493
AY461663.1 - 2 457
BC111088.1 - 2 155 40036837
HY040551.1 - 2 219

View Full Product Details