Gene Transcripts

Gene Symbol
ANO4
Entrez Gene ID
121601
Gene Name
anoctamin 4
Gene Aliases
TMEM16D
Chromosome Location
Chr. 12: 100717457 - 101128641 on Build GRCh38
UniGene ID
Hs.58785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286615.1 NP_001273544.1 28 4159
NM_001286616.1 NP_001273545.1 27 3731
NM_178826.3 NP_849148.2 27 4054
XM_011537911.2 XP_011536213.1 30 4154
XM_011537912.2 XP_011536214.1 30 4158
XM_011537913.2 XP_011536215.1 29 4109
XM_011537914.2 XP_011536216.1 27 3665
XM_011537915.2 XP_011536217.1 28 3977
XM_011537916.2 XP_011536218.1 28 3818
XM_017018809.1 XP_016874298.1 29 4245
XM_017018811.1 XP_016874300.1 26 3862
XM_017018812.1 XP_016874301.1 26 5880
XM_017018813.1 XP_016874302.1 25 3730
XM_017018814.1 XP_016874303.1 28 6119
XM_017018816.1 XP_016874305.1 28 4202
GenBank mRNA AK091540.1 - 18 2675
AK091591.1 - 27 3843

View Full Product Details