Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001251877.1 | NP_001238806.1 | 3 | 384 | |
NM_003363.3 | NP_003354.2 | 3 | 384 | ||
NM_199443.2 | NP_955475.1 | 3 | 384 | ||
GenBank mRNA | AF017305.1 | - | 3 | 308 | |
AF017306.1 | - | 3 | 305 | ||
AK222725.1 | - | 3 | 314 | ||
AK223292.1 | - | 3 | 314 | ||
AK291795.1 | - | 3 | 329 | ||
BC125130.1 | - | 3 | 355 | 40119090 | |
BC125131.1 | - | 3 | 352 | 40119091 | |
BG717217.1 | - | 3 | 389 | ||
U20657.1 | - | 3 | 308 |