Gene Transcripts

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
Chromosome Location
Chr. 10: 59245933 - 59363186 on Build GRCh38
UniGene ID
Hs.607594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001971.2 NP_001001971.1 2 238
NM_001143773.1 NP_001137245.1 3 409
NM_001166698.1 NP_001160170.1 3 323
NM_198215.3 NP_937858.2 2 238
XM_005269618.4 XP_005269675.2 2 238
XM_005269619.4 XP_005269676.2 2 238
XM_006717702.3 XP_006717765.1 4 239
XM_006717703.3 XP_006717766.1 4 421
XM_011539490.2 XP_011537792.1 2 205
XM_011539491.2 XP_011537793.1 3 251
XM_017015885.1 XP_016871374.1 3 152
XM_017015886.1 XP_016871375.1 3 409
XM_017015887.1 XP_016871376.1 3 159
XM_017015888.1 XP_016871377.1 4 421
XM_017015889.1 XP_016871378.1 4 239
XM_017015890.1 XP_016871379.1 3 389
XM_017015891.1 XP_016871380.1 3 409
XM_017015892.1 XP_016871381.1 3 409
XM_017015893.1 XP_016871382.1 4 421
GenBank mRNA AK294805.1 - 2 153
AK299924.1 - 3 409
AK316542.1 - 3 319
BC036453.1 - 2 196 5270033
BC064431.1 - 2 238 5764753
DA188937.1 - 2 238

View Full Product Details