Gene Transcripts

Gene Symbol
NANOS1
Entrez Gene ID
340719
Gene Name
nanos C2HC-type zinc finger 1
Gene Aliases
EC_Rep1a, NOS-1, NOS1, SPGF12, ZC2HC12A
Chromosome Location
Chr. 10: 119029716 - 119033732 on Build GRCh38
UniGene ID
Hs.591918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199461.3 NP_955631.1 1 1349
GenBank mRNA AF275269.2 - 1 1305
AF458985.1 - 1 1349

View Full Product Details