Gene Transcripts

Gene Symbol
NAV1
Entrez Gene ID
89796
Gene Name
neuron navigator 1
Gene Aliases
POMFIL3, STEERIN1, UNC53H1
Chromosome Location
Chr. 1: 201648322 - 201826974 on Build GRCh38
UniGene ID
Hs.585374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020443.4 NP_065176.3 1 936
XM_011510097.1 XP_011508399.1 1 615
XM_011510098.1 XP_011508400.1 1 615
XM_011510099.1 XP_011508401.1 1 615
XM_011510100.1 XP_011508402.1 1 615
XM_011510101.1 XP_011508403.1 1 615
XM_011510102.1 XP_011508404.1 1 615
XM_017002751.1 XP_016858240.1 1 615
XM_017002753.1 XP_016858242.1 1 615
XM_017002754.1 XP_016858243.1 1 615
GenBank mRNA AB032977.3 - 1 269
AJ488101.1 - 1 467
AY043013.1 - 1 936

View Full Product Details