Gene Transcripts

Gene Symbol
FBRS
Entrez Gene ID
64319
Gene Name
fibrosin
Gene Aliases
FBS, FBS1
Chromosome Location
Chr. 16: 30658142 - 30670810 on Build GRCh38
UniGene ID
Hs.247186
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105079.2 NP_001098549.2 18 3112
XM_011545916.2 XP_011544218.1 17 4459
XM_011545917.2 XP_011544219.1 17 4459
XM_011545919.2 XP_011544221.1 16 4422
GenBank mRNA AK022551.1 - 9 1517
AK093045.1 - 2 1788
AK096322.1 - 10 1895
AK298229.1 - 11 1675

View Full Product Details