Gene Transcripts

Gene Symbol
DCLRE1B
Entrez Gene ID
64858
Gene Name
DNA cross-link repair 1B
Gene Aliases
APOLLO, SNM1B, SNMIB
Chromosome Location
Chr. 1: 113904619 - 113914086 on Build GRCh38
UniGene ID
Hs.591412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022836.3 NP_073747.1 2, 3 643
GenBank mRNA AF306692.1 - 1, 2 18
AF306693.1 - 2, 3 578
AK022872.1 - 2, 3 189
AK024060.1 - 2, 3 561
BC029687.1 - 2, 3 520 5001181

View Full Product Details