Gene Transcripts

Gene Symbol
HSPBAP1
Entrez Gene ID
79663
Gene Name
HSPB1 associated protein 1
Gene Aliases
PASS1
Chromosome Location
Chr. 3: 122739997 - 122793829 on Build GRCh38
UniGene ID
Hs.29169
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320728.1 NP_001307657.1 2 321
NM_024610.5 NP_078886.2 2 313
XM_005247762.2 XP_005247819.1 2 313
XM_011513130.2 XP_011511432.1 3 231
GenBank mRNA AF400663.1 - 2 280
AK096705.1 - 2 208
AK289410.1 - 2 313
BC011897.2 - 2 297 3949150
BC063629.1 - 2 253
BI912690.1 - 2 293
HY020808.1 - 2 324

View Full Product Details