Gene Transcripts

Gene Symbol
UBE2I
Entrez Gene ID
7329
Gene Name
ubiquitin conjugating enzyme E2 I
Gene Aliases
C358B7.1, P18, UBC9
Chromosome Location
Chr. 16: 1309153 - 1327018 on Build GRCh38
UniGene ID
Hs.302903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003345.4 NP_003336.1 7 674
NM_194259.2 NP_919235.1 8 931
NM_194260.2 NP_919236.1 7 794
NM_194261.2 NP_919237.1 7 646
XM_017023640.1 XP_016879129.1 7 1071
GenBank mRNA AB208988.1 - 7 755
AJ002385.1 - 7 594
AK291118.1 - 7 665
BC000427.2 - 7 578 2819764
BC004429.1 - 7 578 2819764
BC051289.1 - 7 608
BQ230918.1 - 2 164
BU146357.1 - 3 247
BU543455.1 - 7 666
CD249028.1 - 7 615
CX788607.1 - 7 750
D45050.1 - 7 633
U29092.1 - 6 540
U31882.1 - 7 609
U31933.1 - 7 634
U38785.1 - 7 608
U45328.1 - 7 639
U66818.1 - 7 603
U66867.1 - 7 1322
X96427.1 - 7 639

View Full Product Details