Gene Transcripts

Gene Symbol
ANKRD10
Entrez Gene ID
55608
Gene Name
ankyrin repeat domain 10
Gene Aliases
Chromosome Location
Chr. 13: 110878540 - 110915107 on Build GRCh38
UniGene ID
Hs.525163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017664.3 NP_060134.2 6 1493
NR_104587.1 7 1555
GenBank mRNA AK000100.1 - 6 1455
AK000111.1 - 1 786
AK026017.1 - 2 567
AK094222.1 - 4 1265
AK095763.1 - 4 1165
AK096083.1 - 3 863
AK123341.1 - 1 1328
AK125100.1 - 1 2461
BX641022.1 - 1 2489

View Full Product Details