Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018009.4 | NP_060479.3 | 5 | 1395 | |
XM_005253700.3 | XP_005253757.1 | 5 | 1395 | ||
XM_017019545.1 | XP_016875034.1 | 5 | 1543 | ||
XM_017019546.1 | XP_016875035.1 | 5 | 1223 | ||
GenBank mRNA | AK001005.1 | - | 5 | 1299 | |
AK002056.2 | - | 5 | 1283 | ||
AK098035.1 | - | 6 | 1357 | ||
AK222956.1 | - | 5 | 1283 | ||
AK223163.1 | - | 5 | 1313 | ||
BC015017.2 | - | 5 | 1322 | 3916213 |