Gene Transcripts

Gene Symbol
HIST1H2AL
Entrez Gene ID
8332
Gene Name
histone cluster 1, H2al
Gene Aliases
H2A.i, H2A/i, H2AFI, dJ193B12.9
Chromosome Location
Chr. 6: 27865329 - 27865798 on Build GRCh38
UniGene ID
Hs.233568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003511.2 NP_003502.1 1 412
GenBank mRNA BC069306.1 - 1 412
BC112072.1 - 1 485 8327540

View Full Product Details