Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128429.2 | NP_001121901.1 | 5 | 837 | |
NM_001128430.1 | NP_001121902.1 | 5 | 739 | ||
NM_020159.4 | NP_064544.2 | 5 | 837 | ||
NR_045644.1 | 6 | 888 | |||
XM_017008463.1 | XP_016863952.1 | 5 | 661 | ||
XM_017008464.1 | XP_016863953.1 | 5 | 661 | ||
XM_017008465.1 | XP_016863954.1 | 5 | 671 | ||
GenBank mRNA | AB032948.2 | - | 4 | 625 | |
AK027490.1 | - | 5 | 735 | ||
AY008271.1 | - | 5 | 640 | ||
BC017953.1 | - | 5 | 722 | ||
BC045534.1 | - | 5 | 792 | 5267669 |