Gene Transcripts

Gene Symbol
TULP4
Entrez Gene ID
56995
Gene Name
tubby like protein 4
Gene Aliases
TUSP
Chromosome Location
Chr. 6: 158232192 - 158511828 on Build GRCh38
UniGene ID
Hs.486993
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007466.2 NP_001007467.1 1 1568
NM_020245.4 NP_064630.2 1 1568
XM_011535946.1 XP_011534248.1 2 2245
XM_017011063.1 XP_016866552.1 3 2394
XM_017011064.1 XP_016866553.1 4 2559
XM_017011065.1 XP_016866554.1 3 2312
XM_017011066.1 XP_016866555.1 2 2202
XM_017011067.1 XP_016866556.1 2 2295
XM_017011068.1 XP_016866557.1 2 2261
XM_017011069.1 XP_016866558.1 3 2397
XM_017011070.1 XP_016866559.1 2 2245
XM_017011071.1 XP_016866560.1 3 2397
GenBank mRNA AB037818.1 - 1 1568
AF087973.1 - 328
AF219946.2 - 1 1568
AF288480.1 - 1 1568
AK304056.1 - 1 783
BC152476.1 - 1 1568

View Full Product Details