Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282588.1 | NP_001269517.1 | 4 | 1088 | |
NM_001282589.1 | NP_001269518.1 | 5 | 1192 | ||
NM_001282590.1 | NP_001269519.1 | 6 | 1264 | ||
NM_001282591.1 | NP_001269520.1 | 5 | 1144 | ||
NM_001282592.1 | NP_001269521.1 | 6 | 1481 | ||
NM_001282593.1 | NP_001269522.1 | 4 | 1040 | ||
NM_001282594.1 | NP_001269523.1 | 6 | 1303 | ||
NM_001282595.1 | NP_001269524.1 | 6 | 1046 | ||
NM_001282596.1 | NP_001269525.1 | 5 | 1160 | ||
NM_021181.4 | NP_067004.3 | 7 | 1585 | ||
XM_011509828.1 | XP_011508130.1 | 7 | 1434 | ||
XM_011509829.1 | XP_011508131.1 | 6 | 1330 | ||
GenBank mRNA | AB027233.1 | - | 7 | 1355 | |
AK292148.1 | - | 7 | 1369 | ||
AK298499.1 | - | 5 | 999 | ||
AK298548.1 | - | 4 | 896 | ||
AK301137.1 | - | 4 | 831 | ||
AK301432.1 | - | 6 | 1089 | ||
AK301438.1 | - | 5 | 933 | ||
AL713801.1 | - | 2 | 2210 | ||
AL834424.1 | - | 6 | 1053 | ||
BC027867.1 | - | 6 | 1247 | 5203638 |