Gene Transcripts
- Gene Symbol
- EML2
- Entrez Gene ID
- 24139
- Gene Name
- echinoderm microtubule associated protein like 2
- Gene Aliases
- ELP70, EMAP-2, EMAP2
- Chromosome Location
- Chr. 19: 45609396 - 45645601 on Build GRCh38
- UniGene ID
- Hs.24178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193268.1 |
NP_001180197.1
|
12, 13 |
1525 |
|
NM_001193269.1 |
NP_001180198.1
|
12, 13 |
1429 |
|
NM_012155.2 |
NP_036287.1
|
9, 10 |
890 |
|
NR_034098.1 |
|
7, 8 |
610 |
|
XM_006723125.2 |
XP_006723188.1
|
12, 13 |
1606 |
|
XM_006723126.2 |
XP_006723189.1
|
12, 13 |
1414 |
|
XM_011526694.2 |
XP_011524996.1
|
11, 12 |
1513 |
|
XM_011526695.2 |
XP_011524997.1
|
11, 12 |
1474 |
|
XM_011526698.2 |
XP_011525000.1
|
9, 10 |
932 |
|
XM_011526700.2 |
XP_011525002.1
|
9, 10 |
878 |
|
XM_011526701.2 |
XP_011525003.1
|
4, 5 |
481 |
|
XM_017026550.1 |
XP_016882039.1
|
8, 9 |
774 |
|
XM_017026551.1 |
XP_016882040.1
|
12, 13 |
1609 |
|
GenBank mRNA |
AB209773.1 |
- |
6, 7 |
626 |
|
AF103939.1 |
- |
9, 10 |
884 |
|
AK026616.1 |
- |
7, 8 |
588 |
|
AK225043.1 |
- |
9, 10 |
876 |
|
AK294668.1 |
- |
12, 13 |
1476 |
|
AK295905.1 |
- |
12, 13 |
1429 |
|
AK296258.1 |
- |
12, 13 |
1358 |
|
AK302961.1 |
- |
12, 13 |
1489 |
|
AK304299.1 |
- |
9, 10 |
874 |
|
AL096717.1 |
- |
9, 10 |
861 |
|
BC029304.1 |
- |
12, 13 |
1592 |
|
BC032630.1 |
- |
9, 10 |
875 |
5402615 |
View Full Product Details