Gene Transcripts

Gene Symbol
FBXL5
Entrez Gene ID
26234
Gene Name
F-box and leucine rich repeat protein 5
Gene Aliases
FBL4, FBL5, FLR1
Chromosome Location
Chr. 4: 15604384 - 15681545 on Build GRCh38
UniGene ID
Hs.643433
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193534.1 NP_001180463.1 4 538
NM_001193535.1 NP_001180464.1 4 487
NM_012161.3 NP_036293.1 4 538
NR_036464.1 5 642
XM_006713959.3 XP_006714022.1 5 634
XM_011513831.2 XP_011512133.1 4 320
XM_011513832.2 XP_011512134.1 4 467
XM_011513833.2 XP_011512135.1 3 278
XM_017008019.1 XP_016863508.1 6 1405
GenBank mRNA AF142481.1 - 4 460
AF157323.1 - 4 399
AF176700.1 - 4 422
AF199420.1 - 4 427
AK000153.1 - 3 248
AK295510.1 - 4 457
AK310860.1 - 4 467
AL832808.1 - 2 174
BC012320.1 - 5 571
BC030656.1 - 4 509 4837080
BI464829.1 - 4 520
BX537957.1 - 4 433

View Full Product Details