Gene Transcripts

Gene Symbol
GOLIM4
Entrez Gene ID
27333
Gene Name
golgi integral membrane protein 4
Gene Aliases
GIMPC, GOLPH4, GPP130, P138
Chromosome Location
Chr. 3: 168008672 - 168095925 on Build GRCh38
UniGene ID
Hs.143600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308155.1 NP_001295084.1 15 3028
NM_014498.4 NP_055313.1 16 3112
XM_005247364.2 XP_005247421.1 16 3114
XM_005247365.2 XP_005247422.1 15 3030
XM_017006183.1 XP_016861672.1 15 2510
GenBank mRNA AU310391.1 - 1 325
BF130574.1 - 48

View Full Product Details