Gene Transcripts

Gene Symbol
PIK3R4
Entrez Gene ID
30849
Gene Name
phosphoinositide-3-kinase regulatory subunit 4
Gene Aliases
VPS15, p150
Chromosome Location
Chr. 3: 130678934 - 130746852 on Build GRCh38
UniGene ID
Hs.149032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014602.2 NP_055417.1 4 1612
GenBank mRNA AK314219.1 - 4 1559
BC017332.2 - 4 1577
BC057239.1 - 4 1593
BC110318.1 - 4 1558 5492292
BC127105.1 - 4 1429 40124707
BC127106.1 - 4 1429 40124708
Y08991.1 - 4 1598

View Full Product Details