Gene Transcripts

Gene Symbol
SUMF2
Entrez Gene ID
25870
Gene Name
sulfatase modifying factor 2
Gene Aliases
pFGE
Chromosome Location
Chr. 7: 56064007 - 56080672 on Build GRCh38
UniGene ID
Hs.279696
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042469.1 NP_001035934.2 2 277
NM_001042470.1 NP_001035935.2 2 277
NM_001130069.2 NP_001123541.1 2 277
NM_015411.2 NP_056226.2 2 277
XM_011515254.2 XP_011513556.1 2 277
XM_017011939.1 XP_016867428.1 2 277
XM_017011940.1 XP_016867429.1 2 494
GenBank mRNA AK075477.1 - 2 236
AK297042.1 - 2 193
AK297793.1 - 2 191
AK298605.1 - 2 217
AK301627.1 - 2 194
AL050037.1 - 2 193
AY323911.1 - 2 189
AY359103.1 - 2 207
BC006159.1 - 2 175
BC015600.2 - 2 174
BC065222.1 - 2 224
BC084539.1 - 2 207 6599080
BC111092.1 - 2 230 40068651
BM423367.1 - 2 179
BP296294.1 - 2 499
CF131206.1 - 2 229
CR936749.1 - 2 212
CR936757.1 - 2 220
CX164099.1 - 2 155
DA130450.1 - 2 246
DA924174.1 - 2 278
DB010869.1 - 2 195
KU178582.1 - 1 102
KU178583.1 - 1 102
KU178584.1 - 1 102
KU178585.1 - 1 102

View Full Product Details