Gene Transcripts

Gene Symbol
CDC40
Entrez Gene ID
51362
Gene Name
cell division cycle 40
Gene Aliases
EHB3, PRP17, PRPF17
Chromosome Location
Chr. 6: 110180421 - 110232220 on Build GRCh38
UniGene ID
Hs.428147
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015891.2 NP_056975.1 15 3718
GenBank mRNA AB208903.1 - 15 3693
AK001426.1 - 4 2380

View Full Product Details