Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135643.1 | NP_001129115.1 | 5 | 615 | |
NM_001135644.1 | NP_001129116.1 | 5 | 783 | ||
NM_016221.3 | NP_057305.1 | 5 | 615 | ||
XM_005268457.1 | XP_005268514.1 | 4 | 509 | ||
XM_011537644.1 | XP_011535946.1 | 6 | 519 | ||
XM_011537645.1 | XP_011535947.1 | 6 | 431 | ||
XM_011537646.1 | XP_011535948.1 | 6 | 652 | ||
XM_011537647.1 | XP_011535949.1 | 6 | 475 | ||
GenBank mRNA | AF195120.1 | - | 5 | 530 | |
AK000299.1 | - | 5 | 534 | ||
AK125973.1 | - | 5 | 783 | ||
AK299517.1 | - | 5 | 658 | ||
AK307757.1 | - | 5 | 554 | ||
AK311693.1 | - | 4 | 507 | ||
BC026323.1 | - | 5 | 540 | 4815273 | |
KU178637.1 | - | 5 | 513 | ||
KU178638.1 | - | 5 | 513 | ||
KU178639.1 | - | 3 | 398 |