Gene Transcripts

Gene Symbol
DEC1
Entrez Gene ID
50514
Gene Name
deleted in esophageal cancer 1
Gene Aliases
CTS9
Chromosome Location
Chr. 9: 115141818 - 115402644 on Build GRCh38
UniGene ID
Hs.148841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017418.2 NP_059114.1 8 1165
GenBank mRNA AB022761.1 - 8 1168

View Full Product Details