Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018094.4 | NP_060564.2 | 1 | 599 | |
GenBank mRNA | AJ251548.1 | - | 1 | 357 | |
AK023155.1 | - | 1 | 543 | ||
AK225671.1 | - | 1 | 564 | ||
AK298868.1 | - | 2 | 293 | ||
BC036077.1 | - | 1 | 553 | 5313878 |