Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019851.2 | NP_062825.1 | 3 | 759 | |
GenBank mRNA | AB030648.1 | - | 3 | 626 | |
AB044277.1 | - | 3 | 759 | ||
BC096720.1 | - | 4 | 554 | ||
BC098128.1 | - | 4 | 555 | ||
BC098153.1 | - | 4 | 555 | ||
BC098339.1 | - | 4 | 555 | ||
BC127125.1 | - | 3 | 424 | ||
BC137446.1 | - | 3 | 697 | 9021446 | |
BC137447.1 | - | 3 | 697 | 9021447 | |
BC141979.1 | - | 3 | 454 | ||
BC141980.1 | - | 4 | 598 |