Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271028.1 | NP_001257957.1 | 2 | 404 | |
NM_021153.3 | NP_066976.1 | 2 | 404 | ||
NR_073130.1 | 2 | 404 | |||
XM_011525931.2 | XP_011524233.1 | 2 | 507 | ||
XM_011525932.1 | XP_011524234.1 | 1 | 182 | ||
XM_017025711.1 | XP_016881200.1 | 2 | 429 | ||
GenBank mRNA | AJ007607.1 | - | 2 | 245 | |
AK304000.1 | - | 2 | 354 | ||
AK315257.1 | - | 2 | 406 | ||
AY358654.1 | - | 1 | 143 | ||
BC015877.1 | - | 2 | 296 | 4716394 |