Gene Transcripts

Gene Symbol
CELF2
Entrez Gene ID
10659
Gene Name
CUGBP, Elav-like family member 2
Gene Aliases
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location
Chr. 10: 10462515 - 11336675 on Build GRCh38
UniGene ID
Hs.309288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025076.2 NP_001020247.1 13 1775
NM_001025077.2 NP_001020248.1 13 1614
NM_001083591.1 NP_001077060.1 13 1769
NM_001326317.1 NP_001313246.1 15 1616
NM_001326318.1 NP_001313247.1 15 1634
NM_001326319.1 NP_001313248.1 17 1726
NM_001326320.1 NP_001313249.1 16 1728
NM_001326321.1 NP_001313250.1 16 1658
NM_001326323.1 NP_001313252.1 17 1770
NM_001326324.1 NP_001313253.1 14 1611
NM_001326325.1 NP_001313254.1 15 1687
NM_001326326.1 NP_001313255.1 14 1612
NM_001326327.1 NP_001313256.1 14 1630
NM_001326328.1 NP_001313257.1 15 1724
NM_001326329.1 NP_001313258.1 13 1646
NM_001326330.1 NP_001313259.1 13 1664
NM_001326331.1 NP_001313260.1 13 1816
NM_001326332.1 NP_001313261.1 13 1798
NM_001326333.1 NP_001313262.1 12 1736
NM_001326334.1 NP_001313263.1 14 2001
NM_001326335.1 NP_001313264.1 13 1810
NM_001326336.1 NP_001313265.1 14 1870
NM_001326337.1 NP_001313266.1 12 1630
NM_001326338.1 NP_001313267.1 12 1704
NM_001326339.1 NP_001313268.1 12 1722
NM_001326340.1 NP_001313269.1 14 1764
NM_001326341.1 NP_001313270.1 13 1710
NM_001326342.1 NP_001313271.1 13 1728
NM_001326343.1 NP_001313272.1 14 1782
NM_001326344.1 NP_001313273.1 13 1500
NM_001326345.1 NP_001313274.1 13 1596
NM_001326346.1 NP_001313275.1 11 1743
NM_001326347.1 NP_001313276.1 13 1920
NM_001326348.1 NP_001313277.1 13 1906
NM_001326349.1 NP_001313278.1 13 1924
NM_006561.3 NP_006552.3 13 1721
XM_005252354.4 XP_005252411.1 13 1811
XM_005252357.4 XP_005252414.1 11 1412
XM_005252358.3 XP_005252415.1 11 1753
XM_006717373.2 XP_006717436.1 12 1851
XM_011519294.2 XP_011517596.1 13 1793
XM_011519297.1 XP_011517599.1 13 1916
XM_017015542.1 XP_016871031.1 12 1499
XM_017015543.1 XP_016871032.1 13 1589
XM_017015544.1 XP_016871033.1 13 1577
XM_017015545.1 XP_016871034.1 13 1571
XM_017015546.1 XP_016871035.1 13 1571
XM_017015547.1 XP_016871036.1 13 1823
XM_017015548.1 XP_016871037.1 13 1559
XM_017015549.1 XP_016871038.1 13 1559
XM_017015550.1 XP_016871039.1 13 2557
XM_017015551.1 XP_016871040.1 13 1805
XM_017015552.1 XP_016871041.1 13 1946
XM_017015553.1 XP_016871042.1 13 1934
XM_017015554.1 XP_016871043.1 13 1916
XM_017015556.1 XP_016871045.1 12 1863
XM_017015557.1 XP_016871046.1 14 1528
XM_017015558.1 XP_016871047.1 12 1515
XM_017015559.1 XP_016871048.1 12 1851
XM_017015560.1 XP_016871049.1 12 1512
XM_017015561.1 XP_016871050.1 12 1839
XM_017015562.1 XP_016871051.1 12 1833
XM_017015563.1 XP_016871052.1 12 1820
XM_017015564.1 XP_016871053.1 11 1439
XM_017015565.1 XP_016871054.1 11 1442
XM_017015566.1 XP_016871055.1 11 1427
XM_017015567.1 XP_016871056.1 12 1567
XM_017015568.1 XP_016871057.1 14 1554
XM_017015569.1 XP_016871058.1 12 1848
XM_017015570.1 XP_016871059.1 11 1424
XM_017015571.1 XP_016871060.1 12 1836
XM_017015572.1 XP_016871061.1 12 1537
GenBank mRNA AF036956.1 - 13 1691
AF090693.1 - 13 1589
AF090694.1 - 13 1559
AF432906.1 - 13 1621
AK293164.1 - 13 1662
AK295511.1 - 11 1362
AK297313.1 - 11 1534
AK299548.1 - 13 1551
AK299992.1 - 13 1690
AK302681.1 - 10 1308
AK316354.1 - 13 1521
BC036391.1 - 14 1729 5260623
CR936676.1 - 2 1831
CR976278.1 - 3 362
U69546.1 - 13 1508

View Full Product Details