Gene Transcripts

Gene Symbol
FRMPD1
Entrez Gene ID
22844
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
FRMD2
Chromosome Location
Chr. 9: 37602979 - 37746904 on Build GRCh38
UniGene ID
Hs.163990
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014907.2 NP_055722.2 13 1276
XM_011517803.2 XP_011516105.1 14 1678
XM_011517804.2 XP_011516106.1 13 1597
XM_011517805.2 XP_011516107.1 10 1049
XM_011517806.2 XP_011516108.1 9 2285
XM_017014480.1 XP_016869969.1 13 1957
XM_017014481.1 XP_016869970.1 9 1652
XM_017014482.1 XP_016869971.1 10 1650
GenBank mRNA AB023184.1 - 13 1276
AK302739.1 - 10 1249
AK302854.1 - 9 862
BC114965.1 - 13 1255 40082639
BX648967.1 - 13 1826

View Full Product Details