Gene Transcripts

Gene Symbol
DOLK
Entrez Gene ID
22845
Gene Name
dolichol kinase
Gene Aliases
CDG1M, DK, DK1, SEC59, TMEM15
Chromosome Location
Chr. 9: 128945530 - 128947733 on Build GRCh38
UniGene ID
Hs.531563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014908.3 NP_055723.1 2199
GenBank mRNA AK225402.1 - 1484
BC035556.1 - 2052 5224439

View Full Product Details